Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551602 Supplier Invitrogen™ Supplier No. PA551602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IYKIPRASGT QLAAPRDALE VYDVPPTALR VPSSGPYDCP ASFSHPLTRV APQPPGEDDA PYDVPLTPKP PAELEPDLEW EGGREPGPPI YAAPSNLKRA SALLNLYEAP EELLADGEGG GTDEGIYDVP LLGPEAPPSP EPPGALAS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 78%.

Docking protein which plays a central coordinating role for tyrosine-kinase-based signaling related to cell adhesion. May serve as an activator of SRC and a downstream effector. Interacts with the SH3 domain of FYN and with CRK, SRC, and YES.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFS
Gene Accession No. O43281
Gene Alias Cas scaffolding protein family member 3; CAS3; CASS3; EFS; EFS1; EFS2; embryonal Fyn-associated substrate; hEFS; signal transduction protein (SH3 containing); signal-integrating protein; Sin; SRC-interacting protein
Gene Symbols EFS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EFS. Recombinant protein control fragment (Product #RP-89000).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10278
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.