Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EFTUD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318400
This item is not returnable.
View return policy
Description
EFTUD1 Polyclonal antibody specifically detects EFTUD1 in Human samples. It is validated for ImmunoprecipitationSpecifications
EFTUD1 | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
79631 | |
Store at 4°C. Do not freeze. | |
IgG |
Immunoprecipitation | |
1 mg/ml | |
Immunoprecipitation 15-20 ug/mg lysate | |
EFL1, Elongation Factor Tu GTP Binding Domain Containing 1, Elongation Factor Tu GTP-Binding Domain-Containing Protein 1, Elongation Factor-Like 1, FAM42A, HsT19294, Protein FAM42A, RIA1, Ribosome Assembly 1 Homolog, Ribosome Assembly 1 Homolog (Yeast) | |
Recognizes region between residues Between between 50 and 100.. | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction