Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFTUD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562590 Supplier Invitrogen™ Supplier No. PA562590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTLADCLISS NGIISSHLAG KLRYMD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

EFTUD1 is involved in the biogenesis of the 60S ribosomal subunit and translational activation of ribosomes. Together with SBDS, triggers the GTP-dependent release of EIF6 from 60S pre-ribosomes in the cytoplasm, thereby activating ribosomes for translation competence by allowing 80S ribosome assembly and facilitating EIF6 recycling to the nucleus, where it is required for 60S rRNA processing and nuclear export. Has low intrinsic GTPase activity. GTPase activity is increased by contact with 60S ribosome subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFTUD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Efl1
Gene Accession No. Q7Z2Z2
Gene Alias 4932434J20Rik; 6030468D11Rik; AI451340; AU019507; AU022896; D7Ertd791e; Efl1; Eftud1; elongation factor like GTPase 1; elongation factor Tu GTP binding domain containing 1; elongation factor Tu GTP-binding domain-containing protein 1; Elongation factor-like 1; elongation factor-like GTPase 1; FAM42A; HsT19294; Protein FAM42A; RIA1; ribosome assembly 1 homolog
Gene Symbols Efl1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EFTUD1. Recombinant protein control fragment (Product #RP-99503).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79631
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.