Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EFTUD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10040849
Description
EFTUD2 Polyclonal specifically detects EFTUD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
EFTUD2 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
DKFZp686E24196, elongation factor Tu GTP binding domain containing 2, Elongation factor Tu GTP-binding domain-containing protein 2, FLJ44695, hSNU114, KIAA0031U5-116 kDa, SNRP116, Snrp116,116 kDa U5 small nuclear ribonucleoprotein component, Snu114, SNU114 homolog, U5 snRNP specific protein, 116 kD, U5 snRNP-specific protein, 116 kD, U5 snRNP-specific protein, 116 kDa, U5-116KD | |
Rabbit | |
Affinity Purified | |
RUO | |
9343 | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry 1:200-1:1000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:200-1:1000 | |
Q15029 | |
EFTUD2 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Elongation Factor Tu GTP Binding Domain Containing 2 (U5 snRNP-Specific Protein, 116 kD ) using the numbering given in entry NP_004238.2 (GeneID 9343). | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction