Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFTUD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586004 Supplier Invitrogen™ Supplier No. PA586004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EFTUD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFTUD2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene EFTUD2
Gene Accession No. O08810, Q15029
Gene Alias 116 kDa U5 small nuclear ribonucleoprotein component; 116kDa; Eftud2; elongation factor Tu GTP binding domain containing 2; elongation factor Tu GTP-binding domain-containing protein 2; hSNU114; KIAA0031; MFDGA; MFDM; SNRNP116; Snrp116; Snu114; SNU114 homolog; U5 small nuclear ribonucleoprotein 116 kDa; U5 snRNP-specific protein, 116 kDa; U5-116 kDa; U5-116KD
Gene Symbols EFTUD2
Host Species Rabbit
Immunogen Synthetic peptide within Human Eftud2 aa 1-50.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20624, 287739, 9343
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.