Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFTUD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726113 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6113 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6113 Supplier Proteintech Group Inc Supplier No. 102081AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

10208-1-AP is a rabbit polyclonal antibody raised against the N-terminal of human EFTUD2.

EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.

Specifications

Antigen EFTUD2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene EFTUD2
Gene Accession No. O08810, Q15029
Gene Alias EFTUD2, hSNU114, KIAA0031, Snrp116, Snu114, SNU114 homolog, U5 116 kDa, U5 116KD
Gene Symbols Eftud2
Host Species Rabbit
Immunogen EFTUD2 Fusion Protein Ag0279
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20624, 287739, 9343
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.