Learn More
EFTUD2/SNRP116 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | EFTUD2/SNRP116 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1602
![]() |
Bethyl Laboratories, Inc
IHC00161T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1601
![]() |
Bethyl Laboratories, Inc
IHC00161 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig
EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.Specifications
EFTUD2/SNRP116 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Snrp116, Snu114, U5-116KD, 116 kDa U5 small nuclear ribonucleoprotein component, SNU114 homolog, U5 snRNP specific protein, 116 kD, U5 snRNP-specific protein, 116 kD, U5 snRNP-specific protein, 116 kDa, U5-116 kDa, elongation factor Tu GTP-binding domain-containing protein 2, hSNU114 | |
Between 1 and 50 | |
Primary | |
4° C | |
EFTUD2 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O08810, Q15029 | |
20624, 9343 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.