Learn More
EFTUD2/SNRP116 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300957A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Chicken, Bovine and Orangutan
EFTUD2 is component of the U5 snRNP complex required for pre-mRNA splicing.Specifications
| EFTUD2/SNRP116 | |
| Polyclonal | |
| Unconjugated | |
| EFTUD2 | |
| Snrp116, Snu114, U5-116KD, 116 kDa U5 small nuclear ribonucleoprotein component, SNU114 homolog, U5 snRNP specific protein, 116 kD, U5 snRNP-specific protein, 116 kD, U5 snRNP-specific protein, 116 kDa, U5-116 kDa, elongation factor Tu GTP-binding domain-containing protein 2, hSNU114 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20624, 9343 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O08810, Q15029 | |
| EFTUD2 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.