Learn More
EG-VEGF/PK1 Antibody (MM0043-2L13), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB11093514UV
Catalog No. NBT93514UV
Only null left
Add to Cart
Description
Description
EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
EG-VEGF/PK1
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
PROK1
human recombinant EG-VEGF protein
0.1 mL
Primary
This recognizes human EG-VEGF in western blot. It is highly cross reactive rat and mouse EG-VEGF proteins.
Store at 4C in the dark.
IgG2
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0043-2L13
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1
Mouse
Protein G purified
Cell Cycle and Replication
84432
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.