Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EG-VEGF/PK1 Antibody (MM0043-2L13), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11093514V
Description
EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EG-VEGF/PK1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PROK1 | |
human recombinant EG-VEGF protein | |
0.1 mL | |
Primary | |
This recognizes human EG-VEGF in western blot. It is highly cross reactive rat and mouse EG-VEGF proteins. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0043-2L13 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1 | |
Mouse | |
Protein G purified | |
Cell Cycle and Replication | |
84432 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction