Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EG-VEGF/PK1 Antibody (SR1180), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32237550UL
Description
EG-VEGF/PK1 Monoclonal antibody specifically detects EG-VEGF/PK1 in Human, Mouse samples. It is validated for Western Blot, Flow CytometrySpecifications
EG-VEGF/PK1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry | |
SR1180 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1 | |
A synthesized peptide derived from human EG-VEGF/PK1 (Uniprot #: P58294) | |
50 μg | |
Cell Cycle and Replication | |
84432 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction