Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EG-VEGF/PK1 Rabbit anti-Human, Mouse, Rat, Clone: 1B4G2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31567820UL
Description
EG-VEGF/PK1 Monoclonal antibody specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
EG-VEGF/PK1 | |
Monoclonal | |
Unconjugated | |
EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, prokineticin 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human EG-VEGF/PK1 (P58294). | |
20 μg | |
Cell Cycle and Replication | |
84432 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
1B4G2 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction