Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EG5/KIF11 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | EG5/KIF11 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-9188
|
Bethyl Laboratories, Inc
A301076AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis.Specifications
| EG5/KIF11 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Kinesin-like protein KIF11, Kinesin-related motor protein Eg5, Kinesin-like spindle protein HKSP, Thyroid receptor interacting protein 5, TRIP5, Kines | |
| KIF11 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P52732 | |
| 3832 | |
| Between 1006 and C-term | |
| Primary | |
| 4° C | |
| KIF11 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title