Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eg5 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990133
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0133 100 μL
50-199-0134 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0133 Supplier Rockland Immunochemicals Supplier No. 100401J04
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Eg5 is directed against the human Eg5 protein. A BLAST analysis was used to suggest reactivity with human. Cross-reactivity with Eg5 from other sources have not been determined.

This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis.

Specifications

Antigen Eg5
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 46 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene KIF11
Gene Accession No. P52732
Gene Alias EG5; HKSP; KIF11; Kif8; Kifl1; Kines; kinesin 11; kinesin family member 11; kinesin-like 1; Kinesin-like protein 1; Kinesin-like protein KIF11; Kinesin-like spindle protein HKSP; Kinesin-related motor protein Eg5; KNSL1; MCLMR; Thyroid receptor-interacting protein 5; TR-interacting protein 5; TRIP5; TRIP-5
Gene Symbols KIF11
Host Species Rabbit
Immunogen Eg5 was prepared from whole rabbit serum produced by repeated immunizations with a truncated Eg5 construct expressed in E. coli corresponding to human Eg5 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3832
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.