Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGF Antibody (008), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289582AF488
Description
EGF Monoclonal antibody specifically detects EGF in Human samples. It is validated for ELISASpecifications
EGF | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
008 | |
ELISA | |
beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human EGF (NP_001954.2; Asn 971-Arg 1023) . | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites | |
1950 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction