Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGF Antibody (RM0078-2M13) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018419 0.025 mg
NBP121708 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018419 Supplier Novus Biologicals Supplier No. NBP1217080.025MG

Rat Monoclonal Antibody

EGF Monoclonal specifically detects EGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EGF
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0078-2M13
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), HOMG4, pro-epidermal growth factor, URG
Gene Symbols EGF
Host Species Rat
Immunogen Mouse recombinant EGF
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 1950
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant EGF.
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.