Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGF Mouse anti-Human, Clone: 9A1, Abnova™

Catalog No. 89035442 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-442 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-442 Supplier Abnova Corporation Supplier No. MAB14037

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against recombinant human EGF.

  • High specificity
  • High reactivity

Specifications

Antigen EGF
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 9A1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human EGF.
Dilution ELISA (0.125-0.25 ug/mL) Immunocytochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS.
Gene Accession No. P01133
Gene Alias HOMG4/URG
Gene Symbols EGF
Host Species Mouse
Immunogen Recombinant protein corresponding to human EGF.
Purification Method Protein A affinity purification
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1950
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.