Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGF Mouse anti-Human, Unconjugated, Clone: 7C9.14.6, MilliporeSigma™

Catalog No. MABF22942MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22942MI 25 μg
MABF22941MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22942MI Supplier MilliporeSigma Supplier No. MABF229425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EGF Monoclonal antibody specifically detects EGF in Human samples. It is validated for Immunohistochemistry (Paraffin), Affinity Binding Assay, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7C9.14.6
Conjugate Unconjugated
Gene Accession No. NP_001954
Gene Alias Urogastrone
Gene Symbols EGF
Host Species Mouse
Immunogen A recombinant fragment corresponding to 53 amino acids from the C-terminal region of human Epidermal growth factor.
Molecular Weight of Antigen 133.99 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 7C9.14.6 is a mouse monoclonal antibody that detects human epidermal growth factor. It targets an epitope within 53 amino acids from the C-terminal, extracellular domain region.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.