Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EGFR (aa 746-750 deletion) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8336SP
This item is not returnable.
View return policy
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
EGFR | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1956 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Immunocytochemistry | |
752502 | |
Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P00533 | |
EGFR | |
Human EGFR synthetic peptide CPVAIKTSPKAN Accession # P00533 | |
25 μg | |
Primary | |
Detects human EGFR with the aa 746-750 deletion in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction