Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGFL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565602 Supplier Invitrogen™ Supplier No. PA565602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SELEPECDQCK DGYIGPNCNK CENGYYNFDS ICRKCQCHGH VDPVKTPKIC KPESGECINC LHNTTGFWCE NCLEGYVHDL EGNCIKKEVI LPTPEGSTIL VSNASLTTSV PTPVINSTFT PTTLQTIFSV ST Highest antigen sequence identity to the following orthologs - mouse 86%, rat 84%.

MEGF9 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFL5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MEGF9
Gene Accession No. Q9H1U4
Gene Alias 4933405H16Rik; 9630025C07; EGFL5; EGF-like protein 5; EGF-like-domain, multiple 5; Epidermal growth factor-like protein 5; KIAA0818; Megf9; mKIAA0818; multiple EGF like domains 9; multiple EGF-like domains protein 9; multiple EGF-like-domains 9; multiple epidermal growth factor-like domains protein 9; UNQ671/PRO1305
Gene Symbols MEGF9
Host Species Rabbit
Immunogen Recombinant Human EGFL5. Recombinant protein control fragment (Product #RP-106266).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1955
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.