Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EGFL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113379
Description
EGFL5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
MEGF9 is a protein coding gene.
Specifications
| EGFL5 | |
| Polyclonal | |
| Unconjugated | |
| MEGF9 | |
| 4933405H16Rik; 9630025C07; EGFL5; EGF-like protein 5; EGF-like-domain, multiple 5; Epidermal growth factor-like protein 5; KIAA0818; Megf9; mKIAA0818; multiple EGF like domains 9; multiple EGF-like domains protein 9; multiple EGF-like-domains 9; multiple epidermal growth factor-like domains protein 9; UNQ671/PRO1305 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 1955 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.94 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9H1U4 | |
| MEGF9 | |
| Recombinant Human Multiple epidermal growth factor-like domains protein 9 protein (49-178AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction