Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGFL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551642
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551642 Supplier Invitrogen™ Supplier No. PA551642
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSFNHGICDW KQDREDDFDW NPADRDNAIG FYMAVPALAG HKKDIGRLKL LLPDLQPQSN FCLLFDYRLA GDKVGKLRVF VKNSNNALAW EKTTSEDEKW KTGKIQLYQG TDATKSIIFE AERGKGKTGE IAVDGVLLVS GLCPD Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

The EGFL6 gene encodes epidermal growth factor-like domain-containing protein 6, a member of the EGF-like repeat superfamily known for its roles in development and angiogenesis. EGFL6 is expressed at higher levels during embryonic development and has notable expression in various malignant tumors, highlighting its potential involvement in cancer progression. This secreted protein is pivotal in promoting tumor angiogenesis, influencing cancer cell migration and proliferation without affecting wound healing, making it a significant factor in tumor growth and metastasis. EGFL6 is also expressed in adipose tissues, and its role in adipose differentiation suggests it might link obesity to endometrial cancer risks. The protein impacts cellular activities such as cell adhesion, migration, and proliferation, contributing fundamentally to tumor dynamics. Consequently, EGFL6 is considered an attractive target for antibody-based therapies, with research evaluating its utility in cancer intervention strategies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFL6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EGFL6
Gene Accession No. Q8IUX8
Gene Alias EGF like domain multiple 6; EGF repeat-containing protein 6; EGFL6; EGF-like protein 6; EGF-like-domain, multiple 6; Epidermal growth factor-like protein 6; Maeg; MAM and EGF domain containing; MAM and EGF domains-containing gene protein; PP648; Protein W80; UNQ281/PRO320; W80
Gene Symbols EGFL6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EGFL6. Recombinant protein control fragment (Product #RP-94750).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25975
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.