Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGFL7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106408 Supplier Invitrogen™ Supplier No. PA5106408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EGFL7.

Epidermal growth factor-like protein 7 (EGFL7), also known as VE-statin, MEGF7, Notch4-like protein or Zneu1, is a 29.6 kDa protein produced and secreted by endothelium. The protein is also expressed by some normal and malignant tissues. The main function of EGFL7 is to regulate angiogenesis and vasculogenesis. A high level of EFGL-7 can be found in tissues with active angiogenesis of a variety of disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFL7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EGFL7
Gene Accession No. Q6AZ60, Q9QXT5, Q9UHF1
Gene Alias Cbl20; EGF like domain multiple 7; EGFL7; EGF-like domain 7; EGF-like domain-containing protein 7; EGF-like protein 7; EGF-like-domain, multiple 7; Epidermal growth factor-like protein 7; Estrogen-regulated protein CBL20 20.4kD; Estrogen-regulated protein CBL20, 20.4kD; Megf7; multiple EGF-like domain protein 7; multiple EGF-like domains protein 7; multiple epidermal growth factor-like domain protein 7; multiple epidermal growth factor-like domains protein 7; NEU1; NEU1 protein; NOTCH4-like protein; RP11-251M1.2; UNQ187/PRO1449; Vascular endothelial statin; vascular endothelial-statin; VE-STATIN; Zneu1
Gene Symbols EGFL7
Host Species Rabbit
Immunogen A synthesized peptide derived from human EGFL7(Accession Q9UHF1), corresponding to amino acid residues R70-P120.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245963, 353156, 51162
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.