Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFLAM/Pikachurin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1286 Supplier Bioss Supplier No. BS11346R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in both the retinal photoreceptor ribbon synapse formation and physiological functions of visual perception. Necessary for proper bipolar dendritic tip apposition to the photoreceptor ribbon synapse. Promotes matrix assembly and cell adhesiveness.

Specifications

Antigen EGFLAM/Pikachurin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EGFLAM
Gene Accession No. Q63HQ2
Gene Alias 5930412K08; AGRINL; Agrin-like protein; AGRNL; AU040377; EGF like, fibronectin type III and laminin G domains; Egflam; EGF-like, fibronectin type III and laminin G domains; EGF-like, fibronectin type-III and laminin G-like domain-containing protein; Nectican; PIKA; Pikachurin; RGD1306592
Gene Symbols EGFLAM
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Pikachurin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133584
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.