Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR Antibody (B1D8), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234553MFV610
Description
EGFR Monoclonal antibody specifically detects EGFR in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
EGFR | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse (Negative), Rat (Negative) | |
Purified |
Flow Cytometry | |
B1D8 | |
Flow Cytometry | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Microsomes from A431 cells. | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
1956 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction