Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR Antibody (C225 (Cetuximab)), Janelia Fluor™ 549, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275903JF549
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry.Specifications
EGFR | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EGFR | |
The epidermal growth factor receptor is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. | |
0.1 mL | |
Primary | |
Recognises human EGFR. | |
Store at 4°C in the dark. |
Flow Cytometry, ELISA, Immunohistochemistry | |
C225 (Cetuximab) | |
Flow Cytometry, ELISA, Immunohistochemistry | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Human | |
Protein A purified | |
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
1956.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction