Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR Antibody (DH8.3), PE/Atto594 - Mutant, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250599PEATT594
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry.Specifications
| EGFR | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| EGFR | |
| Synthetic peptide corresponding to the junctional region of the truncated receptor LEEKKGNYVVTDHC,conjugated to keyhole limpet haemcyanin. | |
| 0.1 mL | |
| Primary | |
| Clone DH8.3 of EGF R/ErbB1 antibody is specific for Truncated EGFR which is also called EGFR type III, EGFRvIII or delta-EGFR (characterized by a deletion of 267 amino acids in the extracellular domain, leading to a receptor which is unable to bind ligand yet is constitutively active). Because of its impaired internalisation and degradation, EGFRvIII is known to enhance tumourigenesis. NOTE: Clone DH8.3 DOES NOT cross-react with full length or wildtype EGFR (wtEGFR). | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| DH8.3 | |
| Flow Cytometry | |
| avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
| Mouse | |
| Protein G purified | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
| 1956.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction