Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR Antibody (h-R3 (Nimotuzumab)) - Azide and BSA Free, Novus Biologicals™

Human Monoclonal Antibody
$281.00 - $596.50
Specifications
Clone | h-R3 (Nimotuzumab) |
---|---|
Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence, Block/Neutralize |
Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Blocking Assay, Neutralization |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Block/Neutralize.Specifications
h-R3 (Nimotuzumab) | |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Blocking Assay, Neutralization | |
Unconjugated | |
Human | |
EGFR | |
IgG1 κ | |
Binds to human EGFR domain III at a site distinct from antibody 7A7 (Ab00134) (Talavera et al. 2011) |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunofluorescence, Block/Neutralize | |
Monoclonal | |
Purified | |
Human | |
Raised by humanisation of the murine antibody R3 against EGFR purified from placenta membranes and A431 tumors. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title