Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR Antibody (ICR10) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $487.50
Specifications
Antigen | EGFR |
---|---|
Clone | ICR10 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Western Blot (Negative) |
Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
Description
EGFR Monoclonal specifically detects EGFR in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Western Blot (Negative).Specifications
EGFR | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
RUO | |
P00533 | |
1956 | |
Extracellular domain of Human EGF-receptor from head and neck carcinoma. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ICR10 | |
Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Western Blot (Negative) | |
Monoclonal | |
Rat | |
Mouse, Human | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
EGFR | |
IgG2a | |
Ion exchange chromatography | |
EGFR. It does not react with erb-B2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title