Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR Antibody (Matuzumab) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$280.00 - $596.50
Specifications
Antigen | EGFR |
---|---|
Clone | Matuzumab |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Blocking Assay, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25268525
![]() |
Novus Biologicals
NBP2526850.025MG |
0.025 mg |
Each for $280.00
|
|
|||||
NBP25268502
![]() |
Novus Biologicals
NBP2526850.2MG |
0.2 mg |
Each for $596.50
|
|
|||||
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, CyTOF-ready, Block/Neutralize.Specifications
EGFR | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
EGFR | |
IgG | |
Protein A or G purified | |
Binds to human EGFR. |
Matuzumab | |
Western Blot, Flow Cytometry, Blocking Assay, CyTOF | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Hypoxia, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
1956 | |
Domain III of the extracellular domain of human EGRF. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title