Learn More
Promega EGFR (L858R) Kinase Enzyme System, 1mg
Catalog No. PRV6581 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6581 1 mg
1 options
Catalog No. PRV6581 Supplier Promega Supplier No. V6581
Add to Cart
Add to Cart
Request a Quote
Recombinant human EGFR (L858R) (amino acids 695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EGFR is the receptor for members of the EGF family and is a transmembrane glycoprotein that has tyrosine kinase activity.
  • Easily Screen and Profile EGFR (L858R) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C