Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Mouse anti-Human, Clone: 51CT78.40.5, Abnova™

Catalog No. 89934069 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-069 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-069 Supplier Abnova Corporation Supplier No. MAB12326
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human EGFR.

The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq]

Specifications

Antigen EGFR
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 51CT78.40.5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human EGFR.
Dilution Flow Cytometry (1:10-1:50) Immunohistochemistry (1:50-1:100) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Alias ERBB/ERBB1/HER1/PIG61/mENA
Gene Symbols EGFR
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to human EGFR.
Purification Method Protein G purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1956
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.