Learn More
EGFR, p Tyr1173 Antibody (EM-13), Janelia Fluor™ 646, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP144893JF646
Catalog No. NBP44893646
Only null left
Add to cart
Description
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
EGFR
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
EGFR
Synthetic phospho-peptide covering sequence around tyrosine 1173 of human EGFR.
0.1 mL
Primary
The antibody EM-13 reacts with human EGFR (ErbB1 HER1) phosphorylated on tyrosine 1173.
Store at 4°C in the dark.
IgG1
Flow Cytometry
EM-13
Flow Cytometry
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Mouse
Protein A purified
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases
1956
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.