Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR, p Tyr992 Antibody (EM-12), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP144894AF532
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
EGFR | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
EGFR | |
Synthetic phospho-peptide covering sequence around tyrosine 992 of human EGFR. | |
Primary | |
The antibody EM-12 reacts with human EGFR (ErbB1/ HER1) phosphorylated on tyrosine 992. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
EM-12 | |
Western Blot, Flow Cytometry | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Mouse | |
0.1 mL | |
1956 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction