Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFR Rabbit anti-Human, Clone: SN07-36, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267684
Description
EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
EGFR | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ERBB, ErbB1, ERBB1PIG61, HER1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1 | |
Rabbit | |
Protein A purified | |
Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Cellular Signaling, Growth and Development, Hypoxia, Phospho Specific, Signal Transduction, Tumor Suppressors, Tyrosine Kinases | |
1956 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
SN07-36 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
EGFR | |
Synthetic phospho-peptide surrounding S695 of EGFR | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction