Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Rabbit anti-Human, Unconjugated, Clone: AEF-5, Abnova™

Catalog No. 89339768 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-768 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-768 Supplier Abnova Corporation Supplier No. MAB22030
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

EGFR Monoclonal antibody specifically detects EGFR in Human samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF, Flow Cyt.

Specifications

Antigen EGFR
Applications Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone AEF-5
Conjugate Unconjugated
Description epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian)
Gene EGFR
Gene Alias ERBB, ERBB1, HER1, PIG61, mENA
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human EGFR
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1956
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.