Learn More
Promega EGFR (T790M) Kinase Enzyme System, 10microg
Catalog No. PRV4506 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4506 10 μg
1 options
Catalog No. PRV4506 Supplier Promega Supplier No. V4506
Recombinant human EGFR (T790M) (amino acids 695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EGFR is the receptor for members of the EGF family and is a transmembrane glycoprotein that has tyrosine kinase activity.
  • Easily Screen and Profile EGFR (T790M) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C