Learn More
Promega EGFR (T790M, L858R) Kinase Enzyme System, 10microg
Catalog No. PRV5324 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV5324 10 μg
1 options
Catalog No. PRV5324 Supplier Promega Supplier No. V5324
Recombinant human EGFR (T790M, L858R) (amino acids 695-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. EGFR is receptor for members of the EGF family and a transmembrane glycoprotein that has tyrosine kinase activity.
  • Easily Screen and Profile EGFR (T790M, L858R) Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for convenient bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C