Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFRvIII mutation Mouse anti-Human, Unconjugated, Clone: L8A4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC1126100UG
Description
Anti-EGFRvIII mutation, clone L8A4, Cat. No. MABC1126, is a mouse monoclonal antibody that detects Epidermal Growth Factor Receptor variant III and has been tested for use in ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Specifications
| EGFRvIII mutation | |
| Monoclonal | |
| Please refer to lot specific datasheet. | |
| Purified mouse monoclonal antibody IgG1 in PBS without azide. | |
| EGFR;ERBB;ERBB1;HER1 | |
| KLH-conjugated linear peptide corresponding to the predicted amino acid sequence of 14 amino acids at the fusion junction combined with cultured HC2 20 d2 cells. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Clone L8A4 specifically binds to EGFRvIII, a mutated form of EGFR. | |
| Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
| Purified |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| L8A4 | |
| Unconjugated | |
| Epidermal Growth Factor Receptor vIII | |
| Mouse | |
| ∽145 kDa observed. Uncharacterized bands may be observed in some lysate(s). | |
| 100 μg | |
| Apoptosis & Cancer | |
| NP_005219 | |
| Human | |
| Antibodies | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction