Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGFRvIII mutation Mouse anti-Human, Unconjugated, Clone: L8A4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC112625UG
Description
Anti-EGFRvIII mutation, clone L8A4, Cat. No. MABC1126, is a mouse monoclonal antibody that detects Epidermal Growth Factor Receptor variant III and has been tested for use in ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Specifications
EGFRvIII mutation | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG1 in PBS without azide. | |
EGFR;ERBB;ERBB1;HER1 | |
KLH-conjugated linear peptide corresponding to the predicted amino acid sequence of 14 amino acids at the fusion junction combined with cultured HC2 20 d2 cells. | |
Protein G purified | |
RUO | |
Primary | |
Clone L8A4 specifically binds to EGFRvIII, a mutated form of EGFR. | |
Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Purified |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
L8A4 | |
Unconjugated | |
Epidermal Growth Factor Receptor vIII | |
Mouse | |
∽145 kDa observed. Uncharacterized bands may be observed in some lysate(s). | |
25 μg | |
Apoptosis & Cancer | |
NP_005219 | |
Human | |
Antibodies | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction