Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGFRvIII Recombinant Rabbit Monoclonal Antibody (RM419)
GREENER_CHOICE

Catalog No. PIMA536216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536216 Supplier Invitrogen™ Supplier No. MA536216
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

MA5-36216 reacts to human Epidermal Growth Factor Receptor Variant III. No cross reactivity with wild type EGF receptor.

EGFRvIII is included in receptor tyrosine kinase binding ligands of the EGF family and activating several signaling cascades to convert extracellular cues into appropriate cellular responses. Known ligands include EGF, TGFA/TGF-alpha, amphiregulin, epigen/EPGN, BTC/betacellulin, epiregulin/EREG and HBEGF/heparin-binding EGF. Ligand binding triggers receptor homo- and/or heterodimerization and autophosphorylation on key cytoplasmic residues. The phosphorylated receptor recruits adapter proteins like GRB2 which in turn activates complex downstream signaling cascades. Activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STATs modules. May also activate the NF-kappa-B signaling cascade. Also directly phosphorylates other proteins like RGS16, activating its GTPase activity and probably coupling the EGF receptor signaling to the G protein-coupled receptor signaling. Also phosphorylates MUC1 and increases its interaction with SRC and CTNNB1/beta-catenin. Isoform 2 may act as an antagonist of EGF action.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFRvIII
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone RM419
Concentration 0.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene Egfr
Gene Accession No. P00533
Gene Alias 6720426O10Rik; 9030024J15Rik; A830025P08Rik; AI552599; avian erythroblastic leukemia viral (v-erbB) oncogene homolog; avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; Cntn5; CNTN6; contactin 5; contactin 6; contactin-5; Contactin-6; EGFR; EGFR-related peptide; epidermal growth factor receptor; Epidermal growth factor receptor formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); epidermal growth factor receptor, formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); Erbb; ERBB1; ErbB-1; erb-b2 receptor tyrosine kinase 1; Errb1; Errp; Gm507; HER1; hNB-2; HNB-2s; hNB-3; mENA; mNB-3; Nb-2; NB3; NB-3; neural adhesion molecule; neural recognition molecule NB-2; Neural recognition molecule NB-3; NISBD2; PIG61; Proto-oncogene c-ErbB-1; receptor tyrosine-protein kinase erbB-1; wa2; wa-2; Wa5; waved 2
Gene Symbols Egfr
Host Species Rabbit
Immunogen A peptide corresponding to epidermal growth factor receptor variant III (EGFRvIII) (aa 1-100).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1956
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.