Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGLN1/PHD2 Antibody (2445B) - BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$243.50 - $499.50
Specifications
Antigen | EGLN1/PHD2 |
---|---|
Clone | 2445B |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 - 2 μ/mL, Flow Cytometry 0.25 ug, Immunohistochemistry 5 - 20 μ/mL, Immunocytochemistry/Immunofluorescence 5 - 20 μ/mL, Immunohistochemistry-Paraffin 5 - 20 μ/mL, Flow (Intracellular) 0.25 ug |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP276810SS
![]() |
Novus Biologicals
NBP276810SS |
0.025 mg |
Each for $243.50
|
|
|||||
NBP276810
![]() |
Novus Biologicals
NBP276810 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
EGLN1/PHD2 Monoclonal specifically detects EGLN1/PHD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
EGLN1/PHD2 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Human | |
C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
EGLN1 | |
IgG | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
2445B | |
Western Blot 1 - 2 μ/mL, Flow Cytometry 0.25 ug, Immunohistochemistry 5 - 20 μ/mL, Immunocytochemistry/Immunofluorescence 5 - 20 μ/mL, Immunohistochemistry-Paraffin 5 - 20 μ/mL, Flow (Intracellular) 0.25 ug | |
Monoclonal | |
Rabbit | |
PBS with 0.02% sodium azide | |
54583 | |
Partial synthetic peptide from the N terminal portion of the human EGLN1/PHD2 protein | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title