Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGLN1/PHD2 Antibody (2445B), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB7680H
This item is not returnable.
View return policy
Description
EGLN1/PHD2 Monoclonal antibody specifically detects EGLN1/PHD2 in Human, Rat samples. It is validated for Western Blot, Western Blot, ImmunohistochemistrySpecifications
EGLN1/PHD2 | |
Monoclonal | |
HRP | |
C1orf12, EC 1.14.11.29, EC:1.14.11.291, ECYT3, egl nine homolog 1, egl nine-like protein 1, EGLN1, HALAH, HIF prolyl hydroxylase 2, HIFPH2, HIF-PH2, HIF-prolyl hydroxylase 2, HPH2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2, PNAS-118, PNAS-137, Prolyl hydroxylase domain-containing protein 2, SM20, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human EGLN1/PHD2, Ala2-Phe426, Accession # Q9GZT9 | |
0.1 mL | |
Angiogenesis, Cancer, Chromatin Research, Hypoxia, Immunology, Lipid and Metabolism | |
54583 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
2445B | |
PBS | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction