Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGLN1/PHD2 Antibody (2445D) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP280714
Description
EGLN1/PHD2 Monoclonal specifically detects EGLN1/PHD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EGLN1/PHD2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1 μg/mL, Immunohistochemistry : 3-25 μg/mL, Immunohistochemistry-Paraffin : 3-25 μg/mL | |
C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2445D | |
Unconjugated | |
PBS | |
EGLN1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human EGLN1/PHD2 (amino acids 2-426) [UniProt #Q9QZT9] | |
0.1 mg | |
Cancer, Chromatin Research, Hypoxia | |
54583 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction