Learn More
Description
Specifications
Specifications
| Antigen | EGLN1/PHD2 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 366G/76/3 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | C1orf12, EC 1.14.11.29, EC:1.14.11.291, ECYT3, egl nine homolog 1, egl nine-like protein 1, EGLN1, HALAH, HIF prolyl hydroxylase 2, HIFPH2, HIF-PH2, HIF-prolyl hydroxylase 2, HPH2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2, PNAS-118, PNAS-137, Prolyl hydroxylase domain-containing protein 2, SM20, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
