Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGLN2 Mouse anti-Human, Clone: OTI7H3, lyophilized, TrueMAB™
SDP

Catalog No. 501668959
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-8959 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-8959 Supplier OriGene Supplier No. CF808584
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

The hypoxia inducible factor (HIF) is a transcriptional complex which is involved in oxygen homeostasis. At normal oxygen levels, the alpha subunit of HIF is targeted for degration by prolyl hydroxylation. This gene encodes an enzyme responsible for this posttranslational modification. Multiple alternatively spliced variants, encoding the same protein, have been identified.

Specifications

Antigen EGLN2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7H3
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene EGLN2
Gene Accession No. Q96KS0
Gene Alias EIT6, HIF-PH1, HIFPH1, HPH-1, HPH-3, PHD1
Gene Symbols EGLN2
Host Species Mouse
Immunogen Full length human recombit protein of human EGLN2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112398
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.