Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGLN3/PHD3 Antibody (EG188e/d5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 9 publications
$206.50 - $499.50
Specifications
Antigen | EGLN3/PHD3 |
---|---|
Clone | EG188e/d5 |
Concentration | 1 mg/mL |
Dilution | Western Blot 10 ug/ml, Simple Western 1:1000, Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP130440SS
![]() |
Novus Biologicals
NBP130440SS |
0.05 mL |
Each for $206.50
|
|
|||||
NBP130440
![]() |
Novus Biologicals
NBP130440 |
0.2 mL |
Each for $499.50
|
|
|||||
Description
EGLN3/PHD3 Monoclonal specifically detects EGLN3/PHD3 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EGLN3/PHD3 | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Hypoxia | |
Q9H6Z9 | |
112399 | |
Partial recombinant human PHD3/HIF Prolyl Hydroxylase 3 (UniProt# Q9H6Z9) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
27 kDa |
EG188e/d5 | |
Western Blot 10 ug/ml, Simple Western 1:1000, Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
EC 1.14.11, EC 1.14.11.-, EGL nine (C.elegans) homolog 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein 3 isoform, FLJ21620, HIF prolyl hydroxylase 3, HIF-PH3, HIFPH3 MGC125998, HIF-prolyl hydroxylase 3, HPH-1, HPH-3, Hypoxia-inducible factor prolyl hydroxylase 3, MGC125999, pdh3, PHD3, Prolyl hydroxylase domain-containing protein 3 | |
EGLN3 | |
IgG1 | |
Protein G purified | |
Human, monkey and rat. Immunogen sequence has 98% identity to bovine and mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title