Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGR1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178775AF488
Description
EGR1 Polyclonal antibody specifically detects EGR1 in Human, Mouse samples. It is validated for Western BlotSpecifications
EGR1 | |
Polyclonal | |
Western Blot | |
AT225Transcription factor ETR103, early growth response 1, early growth response protein 1, EGR-1, G0S30, KROX24, KROX-24, Nerve growth factor-induced protein A, NGFI-ATranscription factor Zif268, TIS8, ZIF-268, Zinc finger protein Krox-24, ZNF225Zinc finger protein 225 | |
The immunogen for this product maps to a region between residue 1 and 50 of mouse Early Growth Response Protein 1 using the numbering given in entry NP_031939.1 (GeneID 13653). | |
0.1 mL | |
Angiogenesis, Cancer, Prostate Cancer, Transcription Factors and Regulators | |
1958 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction