Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EGR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583116
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583116 Supplier Invitrogen™ Supplier No. PA583116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IFPEPQSQAF PGSAGTALQY PPPAYPAAKG GFQVPMIPDY LFPQQQGDLG LGTPDQKPFQ GLESRTQQPS LTPLSTIKAF ATQSGSQDLK ALNTSYQSQL I.

The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGR1
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EGR1
Gene Accession No. P18146
Gene Alias A530045N19Rik; AT225; early growth response 1; early growth response protein 1; egr; EGR1; EGR-1; ETR103; G0S30; Krox-1; Krox24; Krox-24; Krox-24 nuclear protein; Nerve growth factor-induced protein A; Ngf1; NGF1-A; Ngfi; NGFIA; NGFI-A; TIS8; transcription factor ETR103; Transcription factor Zif268; Zenk; Zfp-6; Zif268; zif-268; Zinc finger protein 225; zinc finger protein Krox-24; ZNF225
Gene Symbols EGR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EGR1. Recombinant protein control fragment (Product #RP-105819).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1958
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.