Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735995 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5995 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5995 Supplier Proteintech Group Inc Supplier No. 551171AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene belongs to the EGR family of C2H2-type zinc-finger proteins. It is a nuclear protein and functions as a transcriptional regulator. The products of target genes it activates are required for differentitation and mitogenesis. Studies suggest this is a cancer suppresor gene.

Specifications

Antigen EGR1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EGR1
Gene Accession No. P08046, P18146
Gene Alias AT225, early growth response 1, EGR 1, EGR1, G0S30, KROX 24, KROX24, NGFI A, TIS8, Transcription factor ETR103, Transcription factor Zif268, ZIF 268, Zinc finger protein 225, Zinc finger protein Krox 24, ZNF225
Gene Symbols EGR1
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13653, 1958
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.